(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (1R,2R)-(-)-1,2-Bis(methylamino)cyclohexane, (1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine

OR52130-5G OR52130-1G
77410-214EA 23.4 USD
77410-214 77410-212
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 95%
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine
Formula: C₈H₁₈N₂
Boiling Pt: 78…80 °C/18 mmHg
Melting Pt: 36…44 °C
Density: 0.902 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671527
CAS Number: 68737-65-5
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR