Bis(acetonitrile)palladium(II) dichloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Bis(acetonitrile)-palladium(II) chloride, Palladium(II) chloride diacetonitrile complex, Dichlorobis(acetonitrile)palladium(II)

OR46250-1G OR46250-5G
77409-908EA 83.54 USD
77409-908 77409-910
Bis(acetonitrile)palladium(II) dichloride 95%
Bis(acetonitrile)palladium(II) dichloride
Formula: C₄H₆Cl₂N₂Pd
MW: 259.43 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013122
CAS Number: 14592-56-4
EINECS: 238-637-3
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR