m-Toluidine 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Aminotoluene, 3-Toluidine, 3-Methylaniline

OR5362-1KG OR5362-250G OR5362-50G
77410-312EA 144.68 USD
77410-312 77410-314 77410-316
m-Toluidine 99%
m-Toluidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 203
Melting Pt: -30
Density: 0.999 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007808
CAS Number: 108-44-1
EINECS: 203-583-1
UN: 1708
ADR: 6.1 II
Merck Index: 13,09614

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR