2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide 98%

Supplier: APOLLO SCIENTIFIC

OR50999-25G OR50999-5G
77410-142EA 85.58 USD
77410-142 77410-144
2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide 98%
2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide
Formula: C₅H₁₀ClO₃P
MW: 184.558 g/mol
Melting Pt: 97…102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425416
CAS Number: 4090-55-5
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR