2-(Methylthio)ethylamine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Methylthio)ethanamine

OR4873-1G OR4873-25G OR4873-5G
77410-058EA 24.46 USD
77410-058 77410-060 77410-062
2-(Methylthio)ethylamine
2-(Methylthio)ethylamine
Formula: C₃H₉NS
Boiling Pt: 146…149 °C
Density: 0.974 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Freezer
MDL Number: MFCD00014825
CAS Number: 18542-42-2
EINECS: 242-412-5
UN: 2734
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR