4-(Bromomethyl)-3-iodobenzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Cyano-2-iodobenzyl bromide

OR40430-100G OR40430-25G OR40430-5G
77409-536EA 819.12 USD
77409-536 77409-538 77409-540
4-(Bromomethyl)-3-iodobenzonitrile
4-(Bromomethyl)-3-iodobenzonitrile
Formula: C₈H₅BrIN
MW: 321.94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16872046
CAS Number: 182287-63-4
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR