5-Chlorobenzofurazan-4-sulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR3923-1G OR3923-250MG
77409-384EA 82.4 USD
77409-384 77409-386
5-Chlorobenzofurazan-4-sulfonyl chloride 97%
5-Chlorobenzofurazan-4-sulfonyl chloride
Formula: C₆H₂Cl₂N₂O₃S
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00203878
CAS Number: 175203-78-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR