2-Bromo-4'-(dimethylamino)acetophenone 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Dimethylamino)phenacyl bromide, 2-Bromo-1-(4-(dimethylamino)phenyl)ethanone

OR3530-1G OR3530-250MG OR3530-5G
77409-306EA 54 USD
77409-306 77409-308 77409-310
2-Bromo-4'-(dimethylamino)acetophenone 95%
2-Bromo-4'-(dimethylamino)acetophenone
Formula: C₁₀H₁₂BrNO
MW: 242.1124 g/mol
Melting Pt: 89…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06409221
CAS Number: 37904-72-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR