2-Chloro-4-nitrophenyl isothiocyanate 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Chloro-1-isothiocyanato-4-nitrobenzene, 1-Chloro-2-isothiocyanato-5-nitrobenzene

OR33667-1G OR33667-25G OR33667-5G
77409-222EA 28.52 USD
77409-222 77409-224 77409-226
2-Chloro-4-nitrophenyl isothiocyanate 97%
2-Chloro-4-nitrophenyl isothiocyanate
Formula: C₇H₃ClN₂O₂S
MW: 214.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041244
CAS Number: 23165-64-2
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR