(R)-(-)-sec-Butylamine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (R)-(-)-2-Butylamine, (R)-(-)-2-Aminobutane

OR59999-1G OR59999-250MG OR59999-5G
77410-656EA 131.42 USD
77410-656 77410-658 77410-660
(R)-(-)-sec-Butylamine
(R)-(-)-sec-Butylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 63 °C
Density: 0.722 g/cm³ (25 °C)
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00064416
CAS Number: 13250-12-9
UN: 2733
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR