N,N,N',N',N''-Pentamethyldiethylenetriamine
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine
OR55651-100G
OR55651-500G
77410-494EA
21.4
USD
77410-494
77410-496
N,N,N',N',N''-Pentamethyldiethylenetriamine
N,N,N',N',N''-Pentamethyldiethylenetriamine
Formula:
C₉H₂₃N₃ MW: 173.30 g/mol Boiling Pt: 198 °C Melting Pt: –20 °C Density: 0.83 g/cm³ (25 °C) Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014876 CAS Number: 3030-47-5 UN: 2922 ADR: 8+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...