N-(tert-Butoxycarbonyl)-1,5-diaminopentane 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N-Boc-1,5-diaminopentane, tert-Butyl N-(5-Aminopentyl)carbamate, tert-Butyl (5-aminopentyl)carbamate, N-(5-Aminoamyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,5-pentanediamine, N-(5-Aminopentyl)carbamic Acid tert-Butyl Ester, NH2-C5-NH-Boc, N-Boc-1,5-pentanediamine

OR52427-1G OR52427-25G OR52427-5G
77410-242EA 66.22 USD
77410-242 77410-244 77410-246
N-(tert-Butoxycarbonyl)-1,5-diaminopentane 98%
N-(tert-Butoxycarbonyl)-1,5-diaminopentane
Formula: C₁₀H₂₂N₂O₂
MW: 202.30 g/mol
Density: 0.972 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00210020
CAS Number: 51644-96-3
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR