2,1,3-benzoxadiazole-4-sulfonyl chloride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Benzofurazan-4-sulphonyl chloride

OR4939-1G OR4939-5G
77410-076EA 55.02 USD
77410-076 77410-078
2,1,3-benzoxadiazole-4-sulfonyl chloride, Technical Grade
2,1,3-benzoxadiazole-4-sulfonyl chloride
Formula: C₆H₃ClN₂O₃S
MW: 218.611 g/mol
Melting Pt: 77…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00068072
CAS Number: 114322-14-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR