4-(Tributylstannyl)-1,2-dimethylbenzene

Supplier: APOLLO SCIENTIFIC

Synonyms: tributyl-(3,4-dimethylphenyl)stannane

OR399039-1G OR399039-250MG
77409-408EA 303.6 USD
77409-408 77409-410
4-(Tributylstannyl)-1,2-dimethylbenzene
4-(Tributylstannyl)-1,2-dimethylbenzene
Formula: C₂₀H₃₆Sn
MW: 395.21 g/mol
Storage Temperature: Ambient
CAS Number: 1505287-81-9
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR