2,1,3-Benzoxadiazole-5-carbonyl chloride 95+%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-(Chlorocarbonyl)-2,1,3-benzoxadiazole, Benzofurazan-5-carbonyl chloride, 5-(Chloroformyl)-2,1,3-benzoxadiazole

OR3608-1G OR3608-250MG OR3608-5G
77409-314EA 117.16 USD
77409-314 77409-316 77409-318
2,1,3-Benzoxadiazole-5-carbonyl chloride 95+%
2,1,3-Benzoxadiazole-5-carbonyl chloride
Formula: C₇H₃ClN₂O₂
MW: 182.564 g/mol
Melting Pt: 53…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00276978
CAS Number: 126147-86-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR