Ethyl-2-isocyanatobenzoate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Ethoxycarbonyl)phenyl isocyanate

OR322234-1G OR322234-25G OR322234-5G
77409-164EA 49.26 USD
77409-164 77409-166 77409-168
Ethyl-2-isocyanatobenzoate 97%
Ethyl-2-isocyanatobenzoate

When melted, may contain small amount of sediment.

Formula: C₁₀H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00013849
CAS Number: 76393-16-3
UN: 3335
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR