Methyl-2-naphthoate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 2-naphthoate

OR6584-100G OR6584-25G OR6584-5G
77410-870EA 594.98 USD
77410-870 77410-872 77410-874
Methyl-2-naphthoate 97%
Methyl 2-naphthoate
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 141…143 °C/4 mmHg
Melting Pt: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00017345
CAS Number: 2459-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR