2,4-Dimethylimidazole 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR54335-100G OR54335-25G OR54335-5G
77410-368EA 58.08 USD
77410-368 77410-370 77410-372
2,4-Dimethylimidazole 98%
2,4-Dimethylimidazole
Formula: C₅H₈N₂
Boiling Pt: 266 °C/733 mmHg
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00022365
CAS Number: 930-62-1
EINECS: 213-221-4
UN: 3263
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR