3-Acetylpyridine 97+%

Supplier: APOLLO SCIENTIFIC
Danger

OR5046-100G OR5046-25G
77410-110EA 41.78 USD
77410-110 77410-112
3-Acetylpyridine 97+%
3-Acetylpyridine
Formula: C₇H₇NO
Boiling Pt: 218…220 °C
Melting Pt: 41579 °C
Density: 1.102 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR