(Nitromethyl)benzene 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR480613-1G OR480613-25G OR480613-5G
77410-022EA 50.94 USD
77410-022 77410-024 77410-026
(Nitromethyl)benzene 95%
(Nitromethyl)benzene
Formula: C₇H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00053440
CAS Number: 622-42-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR