1-Naphthylamine 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 1-Aminonaphthalene

OR480601-250G OR480601-25G OR480601-100G
77410-014EA 136.52 USD
77410-014 77410-016 77410-012
1-Naphthylamine 98%
1-Naphthylamine
Formula: C₁₀H₉N
MW: 143.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004016
CAS Number: 134-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR