Piperidine-4-sulfonyl chloride, N-CBZ protected, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR4649-1G OR4649-500MG
77409-934EA 93.74 USD
77409-934 77409-936
Piperidine-4-sulfonyl chloride, N-CBZ protected, Technical Grade
Piperidine-4-sulfonyl chloride, N-CBZ protected
Formula: C₁₃H₁₆ClNO₄S
Storage Temperature: Ambient
MDL Number: MFCD05664659
CAS Number: 287953-54-2
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR