Pyridine 99,8%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR41162-1L
OR41162-500ML
OR41162-5L
77409-604EA
41.78
USD
77409-604
77409-606
77409-608
Pyridine 99,8%
Pyridine
Formula:
C₅H₅N Boiling Pt: 115…116 °C Melting Pt: –42 °C Density: 0.978 g/cm³ (25 °C) Flash Pt: 17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...