Ethyl-2-hydroxyisobutyrate 98%

Supplier: APOLLO SCIENTIFIC

OR46219-25G OR46219-5G
77409-898EA 34.64 USD
77409-898 77409-900
Ethyl-2-hydroxyisobutyrate 98%
Ethyl-2-hydroxyisobutyrate
Formula: C₆H₁₂O₃
Boiling Pt: 149…151 °C
Density: 0.965 g/cm³ (25 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00004458
CAS Number: 80-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR