Furfuryl methyl sulfide 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR451317-100G OR451317-25G
77409-802EA 59.1 USD
77409-802 77409-804
Furfuryl methyl sulfide 97+%
Furfuryl methyl sulphide
Formula: C₆H₈OS
Boiling Pt: 64…65 °C
Density: 1.07 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00009604
CAS Number: 1438-91-1
UN: 3334
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR