5-(Bromoacetyl)benzo[b]thiophene 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-(Bromoacetyl)-1-benzothiophene, 1-(Benzo[b]thiophen-5-yl)-2-bromoethan-1-one

OR4407-1G OR4407-250MG
77409-732EA 159.96 USD
77409-732 77409-734
5-(Bromoacetyl)benzo[b]thiophene 97%
5-(Bromoacetyl)benzo[b]thiophene
Formula: C₁₀H₇BrOS
MW: 255.131 g/mol
Boiling Pt: 118…121 °C (20 mmHg)
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD07368508
CAS Number: 1131-87-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR