1-Acetyl-5-indolinesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-acetyl-2,3-dihydro-1H-indole-5-sulphonyl chloride, n-acetylindoline-5-sulphonyl chloride, 1-acetylindoline-5-sulphonyl chloride

OR4396-1G OR4396-250MG OR4396-5G
77409-724EA 57.06 USD
77409-724 77409-726 77409-728
1-Acetyl-5-indolinesulfonyl chloride 97%
1-Acetyl-5-indolinesulfonyl chloride
Formula: C₁₀H₁₀ClNO₃S
Melting Pt: 172…174 °C
Storage Temperature: Freezer
MDL Number: MFCD07368558
CAS Number: 52206-05-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR