2-Iodobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: o-Iodobenzonitrile

OR4224-1G OR4224-25G OR4224-5G
77409-650EA 21.26 USD
77409-650 77409-652 77409-654
2-Iodobenzonitrile 98%
2-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
Melting Pt: 51…55 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079761
CAS Number: 4387-36-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR