3,5-Dibromoanisole

Supplier: APOLLO SCIENTIFIC
Danger

OR42208-1G OR42208-25G OR42208-5G
77409-642EA 21.26 USD
77409-642 77409-644 77409-646
3,5-Dibromoanisole
3,5-Dibromoanisole
Formula: C₇H₆Br₂O
Boiling Pt: 124 °C/9.75 mmHg
Melting Pt: 34…38 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02258848
CAS Number: 74137-36-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR