2-(4-Aminophenyl)ethylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(2-Aminoethyl)aniline, 1-Amino-2-(4-aminophenyl)ethane, 4-Aminophenethylamine

OR40587-10G OR40587-5G
77409-560EA 41.78 USD
77409-560 77409-562
2-(4-Aminophenyl)ethylamine 98%
2-(4-Aminophenyl)ethylamine
Formula: C₈H₁₂N₂
MW: 136.20 g/mol
Boiling Pt: 103 °C/0.3 mmHg
Melting Pt: 28…31 °C
Density: 1.034 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008194
CAS Number: 13472-00-9
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR