1H-Indazole-5-sulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR305122-100MG OR305122-1G OR305122-250MG
77408-882EA 162 USD
77408-882 77408-884 77408-886
1H-Indazole-5-sulfonyl chloride 97%
1H-Indazole-5-sulfonyl chloride
Formula: C₇H₅ClN₂O₂S
MW: 216.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11650650
CAS Number: 599183-35-4
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR