4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR301166-1G OR301166-250MG
77408-800EA 195.62 USD
77408-800 77408-802
4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine 95%
4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine
Formula: C₈H₇BrN₂
MW: 211.0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15529172
CAS Number: 1014613-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR