Piperidine-2,4-dione 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR48245-1G OR48245-25G OR48245-5G
77410-034EA 28.52 USD
77410-034 77410-036 77410-038
Piperidine-2,4-dione 97%
Piperidine-2,4-dione
Formula: C₅H₇NO₂
Melting Pt: 98…103 °C
Storage Temperature: Ambient
MDL Number: MFCD08704814
CAS Number: 50607-30-2
UN: 3335
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR