2-Piperidylacetic acid hydrochloride

Supplier: APOLLO SCIENTIFIC
Danger

OR470052-1G OR470052-5G
77409-960EA 144.68 USD
77409-960 77409-962
2-Piperidylacetic acid hydrochloride
2-Piperidylacetic acid hydrochloride
Formula: C₇H₁₃NO₂·ClH
Melting Pt: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD08061346
CAS Number: 19615-30-6
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR