Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR4200T-1G OR4200T-5G
77409-630EA 452.17 USD
77409-630 77409-632 77409-634
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite 96%
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite

A useful phosphorylating reagent.

Formula: C₁₂H₂₂N₃O₂P
Storage Temperature: Freezer
MDL Number: MFCD00797597
CAS Number: 102690-88-0
UN: 3278
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR