2-Bromo-4-chloro-6-cyclohexylphenol

Supplier: APOLLO SCIENTIFIC
Danger

OR41191-1G OR41191-250MG OR41191-5G
77409-614EA 24.46 USD
77409-614 77409-616 77409-618
2-Bromo-4-chloro-6-cyclohexylphenol
2-Bromo-4-chloro-6-cyclohexylphenol
Formula: C₁₂H₁₄BrClO
MW: 289.596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32111676
CAS Number: 2367002-72-8
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR