4-[(Dimethylamino)methylene]-2,4-dihydro-3H-pyrazol-3-one 95%

Supplier: APOLLO SCIENTIFIC

OR32815-1G OR32815-250MG
77409-202EA 311.76 USD
77409-202 77409-204
4-[(Dimethylamino)methylene]-2,4-dihydro-3H-pyrazol-3-one 95%
4-[(Dimethylamino)methylene]-2,4-dihydro-3H-pyrazol-3-one
Formula: C₆H₉N₃O
MW: 139.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00138484
CAS Number: 821016-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR