2-(Trimethylsilyl)ethoxymethyl chloride 90%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Chloromethoxy)ethyltrimethylsilane, 2-(Trimethylsilyl)ethoxymethyl Chloride, SEM-chloride

OR323097-25G OR323097-100G OR323097-10G
77409-194EA 124.3 USD
77409-194 77409-190 77409-192
2-(Trimethylsilyl)ethoxymethyl chloride 90%
2-(Trimethylsilyl)ethoxymethyl chloride
Formula: C₆H₁₅ClOSi
Boiling Pt: 170…172 °C
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009919
CAS Number: 76513-69-4
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR