4,4-Dimethylcyclohex-2-en-1-on-2-ylboronic acid pinacol ester

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 4,4-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-enone

OR3187-100MG OR3187-50MG
77409-120EA 52.98 USD
77409-120 77409-122
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronic acid pinacol ester
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronic acid pinacol ester
Formula: C₁₄H₂₃BO₃
MW: 250.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027074
CAS Number: 219489-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR