tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: tert-Butyl 3-methyl-4-oxopiperidine-1-carboxylate

OR318070-1G OR318070-5G OR318070-10G
77409-114EA 21.32 USD
77409-114 77409-116 77409-112
tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate 95%
tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate
Formula: C₁₁H₁₉NO₃
MW: 213.2735 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460962
CAS Number: 181269-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR