4-Bromocrotonic acid 95%, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: trans-4-Bromo-2-butenoic Acid, (E)-4-Bromobut-2-enoic acid

OR313099-1G OR313099-25G OR313099-5G
77409-036EA 23.44 USD
77409-036 77409-038 77409-040
4-Bromocrotonic acid 95%, Technical Grade
4-Bromocrotonic acid
Formula: C₄H₅BrO₂
MW: 164.99 g/mol
Melting Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082701
CAS Number: 13991-36-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR