3-Oxetanamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-amino-oxetane, 3-Aminooxetane

OR308008-10G OR308008-1G OR308008-5G
77408-972EA 218.02 USD
77408-972 77408-974 77408-976
3-Oxetanamine 97%
3-Oxetanamine
Formula: C₃H₇NO
MW: 73.09 g/mol
Boiling Pt: 99 °C
Density: 1.05 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Freezer
MDL Number: MFCD08544363
CAS Number: 21635-88-1
UN: 1992
ADR: 3+6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR