1-Methylcyclohexane-1-carboxaldehyde, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Formyl-1-methylcyclohexane

OR30798-10G OR30798-1G OR30798-5G
77408-962EA 828.3 USD
77408-962 77408-964 77408-966
1-Methylcyclohexane-1-carboxaldehyde, Technical Grade
1-Methylcyclohexane-1-carboxaldehyde
Formula: C₈H₁₄O
MW: 126.196 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12028616
CAS Number: 6140-64-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR