Methyl-3-aminobenzoate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 3-aminobenzoate

OR30089-100G OR30089-25G OR30089-500G
77408-708EA 215.98 USD
77408-708 77408-710 77408-712
Methyl-3-aminobenzoate
Methyl-3-aminobenzoate
Formula: C₈H₉NO₂
Boiling Pt: 170 °C/20 mmHg
Melting Pt: 49…55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017102
CAS Number: 4518-10-9
EINECS: 224-842-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR