Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate

Supplier: APOLLO SCIENTIFIC

Synonyms: Methyl 1-Methyl-2-oxopyrrolidine-4-carboxylate

OR300749-1G OR300749-5G
77408-700EA 66.22 USD
77408-700 77408-702
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate
Formula: C₇H₁₁NO₃
MW: 157.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015898
CAS Number: 59857-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR