3-Bromoquinoline-5-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR300389-1G OR300389-250MG
77408-628EA 358.62 USD
77408-628 77408-630
3-Bromoquinoline-5-carboxylic acid
3-Bromoquinoline-5-carboxylic acid
Formula: C₁₀H₆BrNO₂
MW: 252.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20327981
CAS Number: 1344046-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR