7-Bromopyrido[3,4-b]pyrazine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR300382-1G
OR300382-250MG
77408-620EA
338.34
USD
77408-620
77408-622
7-Bromopyrido[3,4-b]pyrazine
7-Bromopyrido[3,4-b]pyrazine
Formula:
C₇H₄BrN₃ MW: 210.031 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22380148 CAS Number: 1337880-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...