5-(Cyclopropyl)thiophene-2-carbonitrile

Supplier: APOLLO SCIENTIFIC

OR45209-1G OR45209-250MG
77409-846EA 660.2 USD
77409-846 77409-848
5-(Cyclopropyl)thiophene-2-carbonitrile
5-(Cyclopropyl)thiophene-2-carbonitrile
Formula: C₈H₇NS
MW: 149.213 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28059614
CAS Number: 1823928-25-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR