1-Amino-1-cyclopropanecarbonitrile hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR40739-1G OR40739-25G OR40739-5G
77409-588EA 24.46 USD
77409-588 77409-590 77409-592
1-Amino-1-cyclopropanecarbonitrile hydrochloride 97%
1-Amino-1-cyclopropanecarbonitrile hydrochloride
Formula: C₄H₇ClN₂
Melting Pt: 214…220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04114063
CAS Number: 127946-77-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR