2,6-Dichloro-3-iodothiophene

Supplier: APOLLO SCIENTIFIC

OR310093-1G OR310093-250MG OR310093-500MG
77409-004EA 246.56 USD
77409-004 77409-006 77409-008
2,6-Dichloro-3-iodothiophene
2,6-Dichloro-3-iodothiophene
Formula: C₄HCl₂IS
Storage Temperature: Ambient
MDL Number: MFCD11844795
CAS Number: 43225-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR